Suspension Clamp
Supplier: United Scientific Supplies
CLHK01
470105-132EA 18.8 USD
470105-132
Suspension Clamp
Clamps General Purpose Clamps
Easy to use hook for your support stand to hold small loads.
  • Aluminum bosshead
  • Rod is 10 cm long
Use to suspend a thermometer over a hot plate or for suspending specific heat specimens. Clamp holds items at a distance of 4 1/2" from the support rod. Aluminum hook swivels and locks at any angle.