Shower Tester
Supplier: ENCON SAFETY PRODUCTS
02-5700-01
470105-050EA 192.5 USD
470105-050
Shower Tester
First Aid Supplies Showers
You know the importance of making sure your safety shower is tested regularly, so it's ready to use in the event of an emergency.
Simply attach this lightweight, PVC coated nylon test funnel to the shower head, and the hemmed bottom passes the water directly and neatly into the bucket.
Will accomodate shower heads up to 8" in diameter. (Bucket not included.)