Boreal Slide Holders with Labels
Supplier: Avantor
470104-916PK 19.9 USD
470104-916
Boreal Slide Holders with Labels
Slide Holders
Excellent for Storing Standard Sized Wholemount Slides
  • Keep slides debris-free
  • Store single slides efficiently
Each crystal-clear holder keeps one slide free of dust. Two internal supports protect the cover glass from pressure. Package includes Boreal slide labels. Package of 20.