Universal Specific Gravity Kit for an Electronic Scale
Supplier: MINERALAB, LLC
SGK-B
470104-828EA 130.79 USD
470104-828
Universal Specific Gravity Kit for an Electronic Scale
Specimens Mineral Specimens Mineral Collections
The Universal Specific Gravity Kit is designed to be used with any electronic scale with a minimum capacity of 15 grams and a tare (re-zeroing) function to easily measure the specific gravity of gemstones and minerals.
Just weigh the stone "in air" on the integrated weighing platform of the scale, then without changing the setup, weigh the stone in water by placing it in the underwater weighing pan of the gem holder suspended above the platform. Plug the two values into the specific gravity equation and obtain the specific gravity of your stone.
Includes: lightweight gem holder support and underwater weighing pan, independent, spill-resistant, adjustable beaker support, beaker for filling with water - 100ml capacity, aluminum test weight, stainless steel tweezers, instructions, reproducible Specific Gravity worksheet, table of 300 common minerals/gems sorted by SG, with 6 other properties, including refractive index.
Note: shown with Boreal Balance - not included.