Earth Science Student Collection
Supplier: Avantor
470104-714EA 232.59 USD
470104-714
Earth Science Student Collection
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
Designed for high school and elementary college courses, these rock and mineral specimens provide a complete range of materials for comparisons of igneous, sedimentary and metamorphic rocks.
  • All specimens are numbered
  • Size of each: 2.5×5 cm
  • Set of 72
In addition, students can study rock-forming ores of metals and non-metals, differences in the hardness scale and physical properties including streak, luster, cleavage, fracture, specific gravity and magnetism. The collection is housed in two 36-compartment collection boxes, one each for rocks and minerals.
Delivery information: Includes streak plate, magnet, magnifier, identification list and instruction booklet.
Caution: Note: Collections are subject to change depending upon availability of individual items. Due to the nature of this product it may contain asbestos.