Stainless Steel Cart with Guard Rails
Supplier: Lakeside
316
470104-692EA 934 USD
470104-692
Stainless Steel Cart with Guard Rails
Carts
A 5 cm high guard-rail on all sides of each shelf prevents small apparatus from falling off this stainless steel laboratory cart.
Clearance between the shelves is 23 cm.
Capacity: 91 kg
Size: 43 x 71 x 84 cm