Adjustable Aluminum Tuning Fork
Supplier: RIVERBANK LABORATORIES INC.
N
470104-596EA 34.75 USD
470104-596
Adjustable Aluminum Tuning Fork
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
This aluminum tuning fork has sliding weights on each tine.
  • High-Quality
  • Adjustable
  • Sliding Weights on Each Tine
Index lines and notes indicate settings from C (128Hz) to B (240Hz).
This high-quality tuning fork is adjustable