Ungraduated Thermometers
Supplier: Echo
BR/2496 PU
470104-552PK 16.9 USD
470104-552
Ungraduated Thermometers
Thermometers Analog Thermometers
Make your own thermometers.
Precision made 12.5" glass tubes with a range of 0 to 245 °F. Filled with a special non-toxic oil that will not fade or separate. Pre-tested for accuracy. Package of six.