Support Ring with Clamp
Supplier: United Scientific Supplies
SRCI03
470104-526EA 16.2 USD
470104-526
Support Ring with Clamp
Clamps General Purpose Clamps
Support rings for funnels of all sizes.
  • Cast iron, zinc plated
  • Boss head clamp
Cast iron, zinc-plated support ring with boss head clamp and thumbscrew. Distance from support to center of ring is 4-1/2" for all sizes.