Replacement Pump Oil
Supplier: Fischer Technical
OIL-012
470104-444CS 450.75 USD
470104-444
Replacement Pump Oil
Lubricants
This is a specially prepared, low vapor pressure mineral oil. It is rust-inhibiting and foam resistant, and permits water condensates to drop out of suspension.