Aluminum Coated Pith-Like Balls
Supplier: Sempco
BH-9107
470104-434PK 19.6 USD
470104-434
Aluminum Coated Pith-Like Balls
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
A thin aluminum coating produces great sensitivity to electrostatic charges. Styrofoam pith-like balls measure 8 mm in diameter and each has a silk suspension cord.
Packaging: Pack of six.