KIMAX® Fermentation Tubes with Foot
Supplier: GSC International
601-9
470104-226EA 15.6 USD
470104-226
KIMAX® Fermentation Tubes with Foot
Tubes Fermentation Tubes
For use in the collection and observation of gases produced by fermentation.
  • Made from borosilicate glass, the bulb is designed to have a capacity greater than that of the vertical tube
  • The vertical tube has a closed, rounded top