Demonstration Balance
Supplier: Avantor
470104-170EA 64.49 USD
470104-170
Demonstration Balance
Educational Classroom Kits and Activities
Inexpensive but Important Apparatus
  • One knife-edge clamp
  • Heavy cast metal support
For demonstrating the principle of moments. A meter stick (not supplied) is easily attached using a set screw fitted on the clamp. Complete with instructions and suggestions for use.