Gas Convection Apparatus
Supplier: CURBELL PLASTICS
6122902
470104-126EA 17.15 USD
470104-126
Gas Convection Apparatus
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
An ideal apparatus for studying convection currents in the air
  • Size: 23 cm x 10 cm x 10 cm
This metal box has a glass front and two plastic chimneys. A candle is placed under one of the chimneys causing the convection currents. A piece of touch paper, producing smoke, is held at the mouth of the other chimney making the convection current dramatically visible.