Boreal Science Blood Typing Kit
Supplier: Avantor
470104-036KT 150.89 USD
470104-036
Boreal Science Blood Typing Kit
Educational Classroom Kits and Activities
Students determine their own blood types with this easy to use kit.
As this kit uses freeze-dried blood typing serum, it does not require refrigeration. Includes materials for 30 students, fliptop box, package of 50 sterile blood lancets, tallquist hemoglobin scale, freeze-dried blood typing serum, instructions.