Sterile Blood Lancet
Supplier: MEDIPOINT, INC.
9810819570
470104-032PK 99.95 USD
470104-032
Sterile Blood Lancet
Blood Lancets
These disposable lancets are a faster, safer, and easier way to take blood samples.
The serrated shape assures a firm finger grip. The sharp, strong point gives a relatively painless puncture. The blood channel provides superior visualization. The lancets are individually packaged, sterilized, and ready for immediate use.
Point Length - 3mm
Total Length of Lancet - 4mm