3B Scientific® Skin
Supplier: American 3B Scientific
1000294
470103-258EA 427.75 USD
470103-258
3B Scientific® Skin
Models Human Biology Models
The model consists of three individual parts on a common stand that represent sections of the human skin with a magnification of 80×. Anatomical differences of the skin sections are didactically represented in an understandable manner.
  • Buy individually or on a single backer
  • Plaque mounted and can be viewed on a table, or hung
  • Three years manufacturers warranty
  • Includes 3B smart anatomy
All layers making up the skin and microscopic structures such as nerve, vessels and tactile corpuscles have been accurately and spatially realized on the models.
Accessories information: For information about 3B smart anatomy, please see the document attached on this page.