Syringe Extender, S17 Science
Supplier: S17 SCIENCE LLC.
EQ 077
470102-726EA 8 USD
470102-726
Syringe Extender, S17 Science
Fittings and Connectors
The extender is used to make an air-tight connection between a syringe and plastic, vinyl, or rubber tubing.