Oven Bake Modeling Clay
Supplier: POLYFORM PRODUCTS COMPANY
S3MP00001
470102-606CS 94.55 USD
470102-606
Oven Bake Modeling Clay
Educational Materials Arts and Crafts Kits and Supplies
Prepare designs easily without premature hardening.
  • Non-toxic
  • Soft and pliable
Can be tooled, rolled, cut, extruded, or formed over molds. To harden, bake in a conventional oven at 275° for 15 minutes. Once cooled, the baked pieces can be polished, drilled, sanded, carved, and painted.
Delivery information: Set contains (10) 2 oz./57 g bars of 10 different colors.
Caution: Conventional oven only. Do not microwave.