Ni-MH Rechargeable Batteries and Chargers
Supplier: RAYOVAC CORPORATION
PL724-4GENE
470039-372CS 196.05 USD
470039-372 470102-430 470102-432 470145-492 470145-490 470145-494 470317-614 470317-640
Ni-MH Rechargeable Batteries and Chargers
Batteries
Spend less on batteries by charging them over and over, and help the envrionment at the same time.
  • Supplied already charged
  • Work in all battery chargers
  • Long lasting and dependable
  • Great for digital devices
Nickel Metal Hydride or NiMH batteries are considered better than NiCad batteries as they are less likely to exhibit the “memory” effect, have a higher storage capacity (+25%), and there is no toxic cadmium to dispose of when they finally wear out. NiMH batteries last an average 6,000 hours and can keep 80% of their charge at -20°. Batteries are rechargeable using the chargers sold separately.