Stoichiometry Survival Kit
Supplier: Avantor
470102-262KT 200.39 USD
470102-262
Stoichiometry Survival Kit
Educational Classroom Kits and Activities
This three part kit uses chalkboard models, manipulatives, and demonstrations to help students gain a better understanding of the math involved with chemistry.
  • Developed by Bernard Uzelac
  • Grades 11 to 12
Mole Map is a flowchart diagram that helps students decide how and when to use chemistry equations. Moles, Molecules, and Atoms is a manipulative that uses removable stickers to help students visualize moles or particles while balancing equations or making conversions between mass, moles, and number of particles. The Dimensional Analysis Blackboard Demo uses laminated magnetic strips, each with a unit printed on it and spaces for the substance and quantity, to show the many problems that can be solved by conversion factors. Includes materials for 15 student groups and instructions.