Rust and Corrosion Kit
Supplier: Avantor
470101-638KT 85.79 USD
470101-638
Rust and Corrosion Kit
Educational Classroom Kits and Activities
This visually striking lab clearly illustrates the processes of rusting, corrosion and cathode inhibition. Iron metal will rust in the presence of air and water, a process that accelerates in ocean water.
  • Developed by Ray Steigvilas
  • Grades 7 to 12
Yet, in the presence of another metal (such as magnesium) the nails will not rust for a very long time as the magnesium sacrifices itself by supplying electrons to the iron nail. Eventually the magnesium will vanish, and a white solid will form on the bottom of the petri dish. Includes materials for five student groups and instructions.