Resonance: The Velocity of Sound Apparatus
Supplier: Avantor
470101-098KT 114.99 USD
470101-098
Resonance: The Velocity of Sound Apparatus
Educational Classroom Kits and Activities
In this kit, ½ wavelengths are measured and used to determine the relationship between wavelength, frequency, and velocity.
  • Developed by physics teacher, Jerome Chandler
  • Grades 9 to 12
  • Length: 122 cm
Simply fill tubes with water, strike a tuning fork above the inside tube, and adjust the inside tube to locate the ½ wavelength nodes. This easy-to-use kit helps students understand the properties of sound waves. Instructions included.
Ordering information: Tuning forks are required but not included.