Pocket Test Thermometers
Supplier: Taylor Precision Products
6079J
470100-978EA 13.85 USD
470100-978
Pocket Test Thermometers
Thermometers Analog Thermometers
Recalibratable thermometers feature a stainless steel thermistor and 2.5cm analog dial.
Thermometers measure from -10 to 110°C with 2° graduations. Includes a protective sheath with clip. Size: 12.5cm long.