NeuLog Anemometer Sensor
Supplier: Wards
NUL242
470100-926EA 320.15 USD
470100-926
NeuLog Anemometer Sensor
Data Loggers
Measure wind velocity.
  • Max. sample rate 100 S/sec.
The NeuLog anemometer sensor enables measurements of wind velocity. Combined with temperature, relative humidity, dew point and barometric pressure sensors, it can be used to make very interesting weather measurements.
Caution: For educational use only.