Large Cylinder and Bottle Brush
Supplier: Allied K & R
E103B
470100-702EA 10.25 USD
470100-702
Large Cylinder and Bottle Brush
Brushes
Reach the bottom of cylinders or bottles with this nylon bristle brush. Made of galvanized wire with soft black bristles, this brush features a special tufted end designed to clean narrow-mouth glassware. The brush area is 7 cm diameter x 20 cm long.