Anesthefly Kit
Supplier: Ward's Science
AA0357-KIT
470100-578EA 47.19 USD
470100-578
Anesthefly Kit
Bottles Drosophila Bottles
A safe, easy method to anesthetize Drosophila.
Flies stay immobilized for at least 50 minutes to several hours without being killed or sterilized.
Delivery information: Includes 100 ml anesthefly solution, cotton-tip swabs, sorting brush and instructions.