Pig Eyes
Supplier: Avantor
470099-652KT 20.95 USD
470099-652
Pig Eyes
Specimens Preserved Specimens Preserved Animal Materials
Formalin Preserved without Color Injection
For dissections, these pig eyes allow students to examine the structures of the eye in detail and to identify eye structures representative of mammals, as they are remarkably similar to the eyes of humans and other mammals.