Eisco® Kidney, Nephron and Glomerulus Model
Supplier: Wards
AM0108
470094-356EA 179.75 USD
470094-356
Eisco® Kidney, Nephron and Glomerulus Model
Models Human Biology Models
Comprehensive kidney structures model.
  • All structures sectioned to reveal internal structures
  • Designed by medical professionals and educators
  • Custom molded using the highest quality nontoxic polymers and PVC
  • Hand-painted by skilled artists
  • Includes a key card
Examine the human kidney in detail with this three part detailed model. A three times life size kidney is bisected to show gross internal structure. A 120× segment showing and nephrons and blood vessels allows for finer examination of the transfer of materials between the kidney and the blood stream. Finally a 700× magnification of a glomerulus allows for detailed discussion of material transfer.