Fat/Lipid Tests
Supplier: Precision Laboratories
LIP-1B-75
470093-682PK 32.65 USD
470093-682
Fat/Lipid Tests
Test Paper Indicator Paper
A simple dip test for presence of lipids in solution.
  • Resealable zip closure bag
  • Waterproof chart
Easily test solutions for the presence of fats with these single use dip tests. Each strip had a single testing pad which will change color in the presence of lipids. Simply dip, wait 2 seconds and compare to the chart.