Protein Test Strips
Supplier: Precision Laboratories
PRO-1V-50
470092-374PK 27.35 USD
470092-374
Protein Test Strips
Test Paper Indicator Paper
Add protein testing to your urinalysis procedures.
  • Flip top vial
  • Waterproof color chart
These single-test test strips have a plastic strip with a reactive pad to prevent contact contamination while handling. Simply dip the pad into the solution, wait 60 seconds, and compare to the color chart on the side of the bottle.