Hematoxylin
Supplier: ALDON CORP SE
CAS Number: 517-28-2
Formula Weight: 302.29
Formula: C16H14O6
Hazard Info: Toxic
Solubility: Water, Alcohol and Glycerin
Synonyms: Natural Black 1, C.I. No. 75290
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...