Plastic Inclined Plane
Supplier: Therm Tech Plastics
295600
470040-322EA 39.15 USD
470040-322
Plastic Inclined Plane
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
This budget-friendly inclined plane provides an excellent track for physics carts.
  • Length: 80 cm
  • Width: 12 cm
  • 1 cm Ø holes for support rod drilled 13 cm from top
This inexpensive, lightweight, and durable channel is a great addition to any physical science or physics classroom. Use it 'upside down' to maximize the useable length (80 cm), or use it with the U-shaped sides facing up to create a track for balls, physics carts, friction blocks and more. This inclined plane is perfect for studying potential/kinetic energy, friction, acceleration, and moment of inertia.
Ordering information: Recommended but not included: ring stand and support rod with boss head. Pulley, carts and more.