Metal Madgeburg Hemispheres
Supplier: United Scientific Supplies
MGH001
470039-952EA 66.15 USD
470039-952
Metal Madgeburg Hemispheres
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Hands on demomstration showing the power of a vacuum.
  • Sturdy metal construction
  • Easy to grab handles
Authentic metal construction makes these heavy duty hemispheres closer to the original version that plastic or rubber replicas. Their thick ircon walls and large steel handles deliver durability for years of student use. A brass plated valve and precision machining also prevent leakage.