Optical Bench, 1.8 m
Supplier: Wards
PH0652
470039-934EA 321.1 USD
470039-934
Optical Bench, 1.8 m
Educational Materials Physics Educational Materials Optics Learning Activities
This is a complete optical bench system at an affordable price.
  • Bright Light Source
  • Lenses and Color Filters Provided
  • Steel Construction
This bench, made from a bent steel U section, is built for years of classroom use.
The set includes everything you need for advanced optical experimentation.
Ordering information: This kit includes a 1.8 m optical bench track with feet supports; 5 pieces of riders; cylindrical lens on holder mounted with slit holder; 5 pieces of lens holder; a 10, 20, and 50 cm convex lenses; a 10, 20, and 50 cm concave lens; set of four color filters with slits; a 100 x 100 mm mirror; a 100 x 100 mm screen; translucent screen; slight holder; light source; and set of diaphragms. A 12V power supply is required but not included.