Earth Force Elementary Education Watershed Field Trip Kit
Supplier: LaMotte
5906
470039-926EA 106.05 USD
470039-926
Earth Force Elementary Education Watershed Field Trip Kit
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
Study the environment and the effect humans have on nature through actual field study.
  • Uses fieldwork to illustrate concepts
  • Includes a teacher's manual
  • Grades 1-5
Study the environment and the effect humans have on nature through actual field study. With this fun, economical kit you will have a hands-on method of explaining a watershed and how it relates to nature that students encounter every day. All necessary reagents and accessories are provided, including tests for pH, nitrate, phosphate, and dissolved oxygen using non-hazardous TesTabs ® ; turbidity tests; temperature tests; and an activity from Project WET. The included teacher's manual provides guidelines on taking a field trip, conducting water quality tests, and data sheets for collection and interpretation of data. Materials are supplied for 30 students. Grades 1-5.