Wind to Hydrogen Science Kit
Supplier: HORIZON FUEL CELL TECHNOLOGIES
FCJJ-56
470039-908EA 394.46 USD
470039-908
Wind to Hydrogen Science Kit
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
The Wind to Hydrogen Science Education Kit enables students to invent their own clean energy applications using a small electric motor powered by a fuel cell.
  • Contains a hydrogen fuel cell and wind turbine
  • Electric motor can be used for various applications
  • 12 years old and up
Hydrogen is produced through water electrolysis by a reversible PEM fuel cell that is itself powered by a wind turbine with profiled blades based on NASA aeronautics. This hydrogen is then converted to electrical energy by the fuel cell and can be used to power a small fan.
Caution: Restriction Comments: This science kit should only be used by persons 12 years old and up, and only under the supervision of adults who have familiarized themselves with the safety measures described in the science kit. Warning: Choking Hazard: Not suitable for children under 3 yrs. The fuel cell generates gases that are very easily ignited.