Autoranging Multimeter
Supplier: NEW GENIE GROUP INC SE
MN36
470039-670EA 75.45 USD
470039-670
Autoranging Multimeter
Electrical Meters Digital Multimeters
With a large, easy-to-read display, this multimeter accurately measures AC and DC voltages to 600V and DC current to 10A
With a large, easy-to-read display, this multimeter accurately measures AC and DC voltages to 600V and DC current to 10A. The thermocoupler temperature measurems to 1400°F (750°C). Features include a resistance test with continuity and diode function and a data hold to lock reading in the display. Includes protective rubber holster, two AAA batteries, test leads, and Type K thermocouple probe.
Certifications: CSA Approved