Demonstration Meter
Supplier: Wards
PH1118E PH1118G PH1116
470039-434EA 47.54 USD
470039-434 470039-436 470039-432
Demonstration Meter
Electrical Meters Voltmeters
This working meter can be used to demonstrate the working principle of AC or DC ammeters and voltmeters with the interchangeable plastic scales, available separately.
Basic sensitivity is 5mA, 100mV. It is housed in a ABS plastic case with glass front and rear. Size: 30 x 15 x 30 cm.
Scale inserts: 0-100 mA (DC); 0-300 V(DC); 0-1A (AC); 0-300 (AC). Package of four.
Scale inserts: 0-1A (DC); -10 to +10 V (DC). Package of two.