Faraday Cage
Supplier: Science First
615-3095
470039-428EA 45.45 USD
470039-428
Faraday Cage
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Demonstrate that a charge cannot exist inside a conductor and re-create Faraday’s classic 'Ice pail' experiment with this kit.
  • With instructions
  • Ø×H: 12×27 cm
Delivery information: Includes a perforated metal cage with stand and cover as well as a mounted metal point for the lightning rod effect.