Normal Anticline and Syncline Model
Supplier: Skywill Far East Industries
805840
470039-170BX 1960.85 USD
470039-170
Normal Anticline and Syncline Model
Models Geology Models
Realistic, detailed rock layers.
  • Key for features is printed on base
  • Displays fold structures in cross-section
  • Shows structural geometry in 3-D
Better than a two-dimensional textbook illustration, this colorful resin model realistically represents a typical anticline/syncline structure. The upright axial plane makes upfold and downfold features easy to identify. The features are labeled and a key is printed on the base.