Economy Dual Magnifier
Supplier: Skywill Far East Industries
120465
470039-164BX 1877.68 USD
470039-164
Economy Dual Magnifier
Magnifiers
The Economy Dual Magnifier is made of unbreakable acrylic.
  • Acrylic Construction
  • Length: 4-1/2"
  • Two Lenses Available
It features two lenses: 3x lens, 5.0 cm in diameter and 6x, one cm in diameter.
The overall length is 4-1/2".