Ward's® Chemistry Acid-Fast Stain Kit
Supplier: Ward's Science
9516700
470038-940EA 38.19 USD
470038-940
Ward's® Chemistry Acid-Fast Stain Kit
Educational Classroom Kits and Activities
Identify the family Mycobacteriaceae by their ability to retain a fuschia color following the staining procedure.
Delivery information: Includes carbol-fuschin solution, methylene blue solution, acid alcohol destain, and instructions.