Confining Clay Layer Mix
Supplier: Avantor
470038-606PK 16.5 USD
470038-606
Confining Clay Layer Mix
Models Stream Models
Mix designed to experiment with confined clay.
  • Mix of fine black sand and pure monmorillonite powder
  • Housed in a 16 oz. jar with screw cap