VWR® Polystyrene Spot Plate
Supplier: CCMI, INC.
2321028
470031-818EA 9 USD
470031-818
VWR® Polystyrene Spot Plate
Staining Tiles
Conduct your microchemistry experiments safely.
  • Chemically inert
  • Unbreakable
Not recommended for use with acetone, xylene, or other strong solvents.