Ward's® Fruit Fly Genetics Demonstration
Supplier: Avantor
470030-324KT 59.69 USD
470030-324
Ward's® Fruit Fly Genetics Demonstration
Educational Classroom Kits and Activities
Demonstrate a simple monohybrid cross using the fruit fly Drosophila melanogaster
  • No microscope needed to view the offspring from this cross
  • Mutations are easily visible
The cross between winged and wingless fruit flies yields a 3:1 winged:wingless ratio.
Males will have white eyes and gray bodies; females will have red eyes and yellow bodies.
All necessary materials are provided, including a hand lens with which students can easily distinguish the trait, as well as males and females.
Kit contains:
Ice pack and anesthetizer to sedate flies
Empty vials and plugs
Instructions and a coupon redeemable for live flies.
Ordering information: This item contains a copupon for live or perishable materials. At checkout, please provide the date you wish the perishable components to arrive.
Delivery information: The kit will ship separately from the live component. Kits do not contain perishable components and generally ship via UPS ground.
Perishable components will ship via overnight or second day delivery.