Ward's® Drosophila Genetics Project Lab Activity
Supplier: Avantor
470030-322KT 269.99 USD
470030-322
Ward's® Drosophila Genetics Project Lab Activity
Educational Classroom Kits and Activities
Perform a variety of investigations utilizing Drosophila melanogaster crosses.
  • Requires only 2 colonies of Drosophila
  • Only pick virgin females once, to teach a wide variety of genetic principals
Teacher developed and tested for years, this lab activity helps you teach monohybrid, dihybrid, and sex-linked crosses; epistasis; crossover; linkage; three-point chromosome mapping; and more.
The set comes with vestigial-sepia and white-miniature-forked cultures as well as a teacher’s guide and student copymasters.
Ordering information: This item contains a coupon for live or perishable materials. At checkout, please provide the date you wish the perishable components to arrive.
Delivery information: The kit will ship separately from the live component. Kits do not contain perishable components and generally ship via UPS ground. Perishable components will ship via overnight or second day delivery.