Ward's® Termite Symbiosis Kit
Supplier: Avantor
470030-314KT 99.09 USD
470030-314
Ward's® Termite Symbiosis Kit
Educational Classroom Kits and Activities
Investigate Mutualism Between a Protist and an Insect
  • Observe a symbiotic relationship
  • Grades 7-12
  • Includes instructions
  • No reproductive termites are sent (we do not send queens)
Students perform a squash of a termite and prepare a wetmount to observe the protist Trichonympha, which resides in the gut of the termite, where it breaks down cellulose. The kit includes termites and WARD’S Internal Guest of Termites video which demonstrates how to prepare the wetmount. Note: Coupon included for perishable materials. Redeem by phone or e-mail. Some states may require a USDA permit.
Ordering information: No reproductive termites are sent with the culture. We do not send queens.