Ward's® Chromatography of Algae Pigments Lab Activity
Supplier: Avantor
470030-206KT 127.79 USD
470030-206
Ward's® Chromatography of Algae Pigments Lab Activity
Educational Classroom Kits and Activities
Examine Algae Pigments Using Thin Layer Chromatography
By separating the pigments from three different algae types that are supplied, students can then analyze the results to study the differences in the pigments used to classify algae types. The set includes: red algae, brown algae, and green algae cultures; a teacher’s manual; student copymaster; and enough materials for six setups. Note: This activity must be performed in a well ventilated area or under a fume hood. Coupon included for perishable materials. Redeem by mail, fax, phone, or e-mail.