BoneClones® Comparative Maxilla Set
Supplier: BONE CLONES HOLDINGS
COMP-105
470030-036KT 270 USD
470030-036
BoneClones® Comparative Maxilla Set
Educational Materials Forensics Educational Materials Forensics Supplies
Excellent Comparative Set For Forensic And Anthropological Studies
Discover forensic methods of identifying the race/ethnicity of an individual with this three-piece set. Differences in the maxillary, nasal, dental and palate structures among Asian, African, and European males are clearly illustrated. In the Asian model, the dental arch is rounded and the lingual surface of the incisors is shovel-shaped; the African replica features a hyperbolic dental arch and spatulate incisors; and the European model’s dental arch is parabolic and the incisors are spatulate.